missing translation for 'onlineSavingsMsg'
Learn More

Quinaldic acid, 98%

Product Code. 11499502
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499502 100 g 100g
11479502 5 g 5g
11489502 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499502 Supplier Thermo Scientific Alfa Aesar Supplier No. A10192.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-10-7 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-10-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006752
InChI Key LOAUVZALPPNFOQ-UHFFFAOYSA-N
Synonym quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
PubChem CID 7124
ChEBI CHEBI:18386
IUPAC Name quinoline-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 156°C to 159°C
Quantity 100 g
Beilstein 126322
Merck Index 14,8046
Formula Weight 173.17
Percent Purity 98%
Chemical Name or Material Quinaldic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.