missing translation for 'onlineSavingsMsg'
Learn More

Quinaldic acid, 99%

Product Code. 10371384 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10371384 50 g Glass bottle 50g
10542771 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371384 Supplier Thermo Scientific Acros Supplier No. 171050500
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 93-10-7 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-10-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00006752
InChI Key LOAUVZALPPNFOQ-UHFFFAOYSA-N
Synonym quinaldic acid, 2-quinolinecarboxylic acid, quinaldinic acid, 2-carboxyquinoline, quinaldate, 2-quinolinylcarboxylic acid, 2-quinoline carboxylic acid, 2-quinolinecarboxylate, 2-quinaldic acid, 2-quinolinecarbocylic acid
PubChem CID 7124
ChEBI CHEBI:18386
IUPAC Name quinoline-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 159.0°C to 161.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Beilstein 22, 71
Merck Index 15, 8162
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alkali solutions,soluble in alcohol
Formula Weight 173.17
Percent Purity 99%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material Quinaldic acid
Show More Show Less
compliance-icons
Product Identifier
  • Quinaldic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.