missing translation for 'onlineSavingsMsg'
Learn More

Quinaldine, 95+%

Product Code. 10582911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10582911 250 mL Glass bottle 250mL
10085683 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10582911 Supplier Thermo Scientific Acros Supplier No. 132172500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 91-63-4 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-63-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006756
InChI Key SMUQFGGVLNAIOZ-UHFFFAOYSA-N
Synonym quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech
PubChem CID 7060
IUPAC Name 2-methylquinoline
SMILES CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point -2.0°C
Color White to Yellow
Density 1.0580g/mL
Boiling Point 248.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6070 to 1.6120
Quantity 250 mL
Beilstein 20, 387
Merck Index 15, 8163
Specific Gravity 1.058
Solubility Information Solubility in water: 2.5g/L (20°C). Other solubilities: soluble in chloroform,ether and alcohol
Formula Weight 143.19
Percent Purity 95+%
Physical Form Crystalline Powder
Chemical Name or Material Quinaldine, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.