missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinaldine Red, dye content, 95%

Product Code. 15442058
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442058 5 g 5g
15452058 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442058 Supplier Thermo Scientific Chemicals Supplier No. H55729.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol

Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).

Solubility
Partly soluble in water, alcohol, and ethanol.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
Synonym quinaldine red, eastman 1361, 2-p-dimethylaminostyryl quinoline ethiodide, 1-ethyl-2-p-dimethylaminostyrylquinoline iodide, 2-4-dimethylaminostyryl-1-ethylquinolinium iodide, 2-4-dimethylamino styryl-1-ethylquinolinium iodide, 2--4-dimethylamino styryl-1-ethylquinolinium iodide, chinaldinrot, alpha-p-dimethylaminophenylethylene quinolineethiodide, quinolinium, 2-2-4-dimethylamino phenyl ethenyl-1-ethyl-, iodide 1:1
PubChem CID 5702759
IUPAC Name 4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

Color Black to Green
Quantity 5 g
Beilstein 3798055
Sensitivity Light sensitive
Merck Index 14,8049
Solubility Information Partly soluble in water,alcohol,and ethanol.
Formula Weight 430.33
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Quinaldine Red, Dye content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.