missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Quinaldine red, pure

Product Code. 10200023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10200023 5 g Glass bottle 5g
10620545 25 g Glass bottle 25g
10638994 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10200023 Supplier Thermo Scientific Chemicals Supplier No. 377510050
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 117-92-0 | C21H23IN2 | 430.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-92-0
Molecular Formula C21H23IN2
Molecular Weight (g/mol) 430.33
MDL Number MFCD00011968
InChI Key JOLANDVPGMEGLK-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)styryl]-1-ethylquinolinium iodide
PubChem CID 5702759
SMILES [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C

Specifications

Infrared Spectrum Authentic
IUPAC Name 2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide
Formula Weight 430.32
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Black/Green or Blue to Gray
Quantity 5 g
Physical Form Powder
Chemical Name or Material Quinaldine red, Pure

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.