missing translation for 'onlineSavingsMsg'
Learn More

Quinidine (>85%%), TRC

Product Code. 30413741
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413741 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413741 Supplier Toronto Research Chemicals Supplier No. TRCQ68507050MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20 H24 N2 O2
Molecular Weight (g/mol) 324.42
Synonym Quinidine Sulfate Imp. A (EP), Quinidine Imp. A (EP), Quinine Dihydrochloride Imp. A (EP), Quinine Imp. A (EP), Quinidine, (+)-Quinidine, (8R,9S)-Quinidine, Chinidin, Cinquin, Conchinin, Conchinine, Conquinine, Kinidin, Pitayin, Pitayine, Quindine, beta-Quinidine, beta-Quinine, Quinine Hydrochloride Impurity A, Quinine Impurity A
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19+,20-/m0/s1
Formula Weight 324.1838
Chemical Name or Material Quinidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.