missing translation for 'onlineSavingsMsg'
Learn More

Quinidine, 98%, anhydrous

Product Code. 10380954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10380954 100 g Glass bottle 100g
10194410 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10380954 Supplier Thermo Scientific Acros Supplier No. 163681000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-54-2 | C20H24N2O2 | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-54-2
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00135581,MFCD00198096
InChI Key LOUPRKONTZGTKE-LHHVKLHASA-N
Synonym quinidine, +-quinidine, chinidin, pitayine, cin-quin, 8r,9s-quinidine, conquinine, conchinin, quinidex, conchinine
PubChem CID 441074
ChEBI CHEBI:28593
SMILES COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1

Specifications

Melting Point 170.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 8176
Specific Rotation + 256.00
Solubility Information Solubility in water: 1g/2000mL (20°C)- 1g/800mL (100°C). Other solubilities: 1g/36mL alcohol- soluble in methanol,1g/56mL ether- 1g/1.6mL chloroform,practically insoluble in petroleum ether
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Formula Weight 324.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Quinidine
Show More Show Less
compliance-icons
Product Identifier
  • Quinidine, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.