missing translation for 'onlineSavingsMsg'
Learn More

Quinidine, TRC

Product Code. 30419511
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419511 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419511 Supplier Toronto Research Chemicals Supplier No. TRCQ68500050MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quinidine
CAS 56-54-2
Molecular Formula C20 H24 N2 O2
Quantity 50 mg
Synonym Quinidine Sulfate Imp. A (EP), Quinidine Imp. A (EP), Quinine Dihydrochloride Imp. A (EP), Quinine Imp. A (EP), Quinidine, (+)-Quinidine, (8R,9S)-Quinidine, Chinidin, Cinquin, Conchinin, Conchinine, Conquinine, Kinidin, Pitayin, Pitayine, Quindine, beta-Quinidine, beta-Quinine, Quinine Hydrochloride Impurity A, Quinine Impurity A
InChI Formula InChI=1S/C20H24N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3/t13-,14-,19+,20-/m0/s1
SMILES COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1
IUPAC Name (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
Molecular Weight (g/mol) 324.42
Formula Weight 324.1838
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.