missing translation for 'onlineSavingsMsg'
Learn More

Quinine, 98+%, anhydrous

Product Code. 10686942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686942 50 g Glass bottle 50g
10367960 10 g Glass bottle 10g
10464795 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10686942 Supplier Thermo Scientific Acros Supplier No. 163700500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130-95-0
Molecular Formula C20H24N2O2
Molecular Weight (g/mol) 324.42
MDL Number MFCD00198096
InChI Key LOUPRKONTZGTKE-ZCXAXGJJNA-N
Synonym 1r-6-methoxyquinolin-4-yl 1s,4s,5r-5-vinylquinuclidin-2-yl methanol, quinine anhydrous, +-2,3-o-isopropylidene-l-threitol
PubChem CID 129316724
IUPAC Name (5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl)-[6-(trideuteriomethoxy)quinolin-4-yl]methanol
SMILES [H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C

Specifications

Melting Point 176.0°C to 177.0°C
Color White
Loss on Drying 1% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 50 g
Fieser 06,501; 07,311; 08,340; 09,403; 10,338; 11,456
Merck Index 15, 8177
Specific Rotation Condition − 172.00 (20.00°C c=1,C2H5OH)
Specific Rotation − 172.00
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkalies and acids (formation of salts),soluble in alcohol,ether,chloroform,glycerol,and carbon disulfide
Formula Weight 324.42
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Quinine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Quinine, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.