missing translation for 'onlineSavingsMsg'
Learn More

Quinocetone, TRC

Product Code. 30413813
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413813 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413813 Supplier Toronto Research Chemicals Supplier No. TRCQ6966001G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81810-66-4
Molecular Formula C18 H14 N2 O3
Molecular Weight (g/mol) 306.32
Synonym 2-Propen-1-one, 1-(3-methyl-2-quinoxalinyl)-3-phenyl-, N,N'-dioxide, 1-(3-Methyl-1,4-dioxido-2-quinoxalinyl)-3-phenyl-2-propen-1-one, NSC 621477, Quinocetone
IUPAC Name (E)-1-(3-methyl-1,4-dioxidoquinoxaline-1,4-diium-2-yl)-3-phenylprop-2-en-1-one
SMILES Cc1c(C(=O)\C=C\c2ccccc2)[n+]([O-])c3ccccc3[n+]1[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H14N2O3/c1-13-18(17(21)12-11-14-7-3-2-4-8-14)20(23)16-10-6-5-9-15(16)19(13)22/h2-12H,1H3/b12-11+
Formula Weight 306.1004
Chemical Name or Material Quinocetone (technical product)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.