missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-4-carboxylic Acid, TRC

Product Code. 30417053
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417053 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417053 Supplier Toronto Research Chemicals Supplier No. TRCQ700508100MG
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Quinoline-4-carboxylic Acid
CAS 486-74-8
Molecular Formula C10H7NO2
Quantity 100 mg
Synonym NSC 13138, 4-Carboxyquinoline, 4-Quinolinic Acid
InChI Formula InChI=1S/C10H7NO2/c12-10(13)8-5-6-11-9-4-2-1-3-7(8)9/h1-6H,(H,12,13)
SMILES OC(=O)c1ccnc2ccccc12
Molecular Weight (g/mol) 173.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.