missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-boronic acid, 96%, Thermo Scientific™

Product Code. 10784723 Shop All Thermo Scientific Products
Change view
Click to view available options
250mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10784723 250mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10784723 Supplier Thermo Scientific Supplier No. 359672500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 86-58-8
CAS Min % 95.0
CAS Max % 100.0
Boiling Point 160°C to 165°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H8BNO2
MDL Number MFCD01114698
Quantity 250mg
Merck Index 15, 8186
Synonym quinoline-8-boronic acid, 8-quinolineboronic acid, 8-quinolinylboronic acid, 8-quinoline boronic acid, quinolin-8-yl boronic acid, 8-boronoquinoline, 8-quinolylboronic acid, boronic acid, 8-quinolinyl, unii-5qs1a25ij6, quinolin-8-yl-8-boronic acid
InChI Key KXJJSKYICDAICD-UHFFFAOYSA-N
SMILES B(C1=C2C(=CC=C1)C=CC=N2)(O)O
IUPAC Name quinolin-8-ylboronic acid
Molecular Weight (g/mol) 172.98
PubChem CID 2734380
Formula Weight 172.98
Percent Purity 96%
Chemical Name or Material Quinoline-8-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.