missing translation for 'onlineSavingsMsg'
Learn More

Quinoline, 96%

Product Code. 11424573
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424573 500 g 500g
11414573 100 g 100g
11434573 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424573 Supplier Thermo Scientific Alfa Aesar Supplier No. A11545.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-22-5 | C9H7N | 129.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Quinoline is used as a chemical intermediate for making medicines and other organic compounds.  It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.

Solubility
More soluble in hot than cold water. Miscible with alcohol, ether

Notes
Dissolves sulfur, As{2}O{3}, phosphorus
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-22-5
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006736,MFCD06637409,MFCD31699982,MFCD31699983,MFCD31699984,MFCD31699985,MFCD31699986,MFCD31699987
InChI Key SMWDFEZZVXVKRB-UHFFFAOYSA-N
Synonym chinolin, chinoline, 1-benzazine, quinolin, 1-azanaphthalene, chinoleine, leucol, benzo b pyridine, benzopyridine, leukol
PubChem CID 7047
ChEBI CHEBI:17362
IUPAC Name quinoline
SMILES C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point -15°C
Density 1.09
pH 7.3
Boiling Point 237°C
Flash Point 101°C (214°F)
Refractive Index 1.626
Quantity 500 g
UN Number UN2656
Beilstein 107477
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,8068
Solubility Information More soluble in hot than cold water; Miscible with alcohol, ether
Formula Weight 129.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Quinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.