Learn More
Quinoline, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Quinoline is used as a chemical intermediate for making medicines and other organic compounds. It acts as a fundamental structure in some antihypertensive agents such as prazosin and doxazosin which are peripheral vasodilator. It is also used in alkaloids, dyes, rubber chemicals and flavoring agents. They are also used as catalyst, corrosion inhibitor, preservative, and as solvent for resins and terpenes. They are used in transition-metal complex catalyst chemistry for uniform polymerization and luminescence chemistry. They are used as antifoaming agent in refinery field.
Solubility
More soluble in hot than cold water. Miscible with alcohol, ether
Notes
Dissolves sulfur, As{2}O{3}, phosphorus
Specifications
Specifications
| Melting Point | -15°C |
| Density | 1.09 |
| pH | 7.3 |
| Boiling Point | 237°C |
| Flash Point | 101°C (214°F) |
| Refractive Index | 1.626 |
| Quantity | 2500 g |
| UN Number | UN2656 |
| Beilstein | 107477 |
| Sensitivity | Hygroscopic; Light sensitive |
| Mostrar más |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.