missing translation for 'onlineSavingsMsg'
Learn More

Quinoline Yellow (Technical Grade), TRC

Product Code. 30004294
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004294 2.5 g 2.5g
30004909 250 mg 250mg
30004433 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004294 Supplier Toronto Research Chemicals Supplier No. TRCQ9900802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 478.4
Synonym C.I. Acid Yellow 3, 1,3-Isobenzofurandione, reaction products with methylquinoline and quinoline, sulfonated, Acid Yellow 3, Acid Yellow QS, Basacid Yellow 094, Basovit Yellow 095E, C.I. 47005, C.I. Food Yellow 13, Cogilor Yellow 112.12, D & C Yellow 10, D and C Yellow No. 10, D&C Yellow No. 10, D&C Yellow No. 10-307007, D&C Yellow No. 10-38009, D&C Yellow No. 10-38012, D&C Yellow No. 10-90125, D&C Yellow No. 10HWS307010, D&C Yellow No. 10W074, Dye Quinoline Yellow, E 104, E 104 (dye), Eurocert Quinoline Yellow 311744, FD and C Yellow No. 10, FD&C Yellow No. 10, Food Yellow No. 13, Japan Yellow 203, Lemon Yellow ZN 3, Neelicol Quinoline Yellow WS, Quinidine Yellow KT, Quinoline Yellow, Quinoline Yellow 36012, Quinoline Yellow 38009, Quinoline Yellow 90125, Quinoline Yellow 90145, Quinoline Yellow Certifiable 307006, Quinoline Yellow E 104, Quinoline Yellow Extra, Quinoline Yellow Lake, Quinoline Yellow S, Quinoline Yellow WG-G, Quinoline Yellow WG-G conc, Quinoline Yellow WS, Quinolone Yellow Extra, Vitasyn Quinoline Yellow 70, Yellow 10, Yellow No. 10, Yellow No. 203
IUPAC Name quinoline yellow
SMILES O=C(C1C2=NC3=C(S(=O)(O[Na])=O)C=C(S(=O)(O[Na])=O)C=C3C=C2)C4=CC=CC=C4C1=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C18H11NO8S2.2Na/c20-17-11-3-1-2-4-12(11)18(21)15(17)13-6-5-9-7-10(28(22,23)24)8-14(16(9)19-13)29(25,26)27;;/h1-8,15H,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2
Formula Weight 476.96
Chemical Name or Material Quinoline Yellow (E104)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.