missing translation for 'onlineSavingsMsg'
Learn More

Quinolinic acid, MedChemExpress

Product Code. 16460699
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460699 Supplier Medchem Express Supplier No. HY100807

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quinolinic acid is an endogenous N-methyl-D-aspartate (NMDA) receptor agonist synthesized from L-tryptophan via the kynurenine pathway and thereby has the potential of mediating N-methyl-D-aspartate neuronal damage and dysfunction.

Quinolinic acid is an endogenous N-methyl-D-aspartate (NMDA) receptor agonist synthesized from L-tryptophan via the kynurenine pathway and thereby has the potential of mediating N-methyl-D-aspartate neuronal damage and dysfunction.

Specifications

Chemical Name or Material Quinolinic acid
CAS 89-00-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H5NO4
Solubility Information DMSO : 33.33 mg/mL (199.44 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (19.93 mM; Need ultrasonic)
SMILES O=C(C1=NC=CC=C1C(O)=O)O
Molecular Weight (g/mol) 167.12
Quantity 1 g
Formula Weight 167.12
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.