missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, 98+%

Product Code. 11497966
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497966 500 g 500g
11487966 100 g 100g
11407976 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497966 Supplier Thermo Scientific Alfa Aesar Supplier No. A14055.36
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-19-0 | C8H6N2 | 130.15 g/mol

Used as dyes, pharmaceuticals and antibiotics intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-19-0
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006719
InChI Key XSCHRSMBECNVNS-UHFFFAOYSA-N
Synonym benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
PubChem CID 7045
ChEBI CHEBI:36616
IUPAC Name quinoxaline
SMILES C1=CC=C2C(=C1)N=CC=N2

Specifications

Melting Point 27°C to 32°C
Density 1.124
Boiling Point 220°C to 223°C
Flash Point 98°C (208°F)
Quantity 500 g
Beilstein 109351
Merck Index 14,8078
Formula Weight 130.15
Percent Purity ≥98%
Physical Form Crystals or Powder or Crystalline Powder or Fused Solid or Needles or Liquid
Chemical Name or Material Quinoxaline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.