missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, 98+%, Thermo Scientific Chemicals

Product Code. 10215330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10215330 100 g Glass bottle 100g
10508893 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215330 Supplier Thermo Scientific Acros Supplier No. 132221000
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-19-0 | Molecular Formula: C8H6N2 | Molecular Weight: 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-19-0
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006719
InChI Key XSCHRSMBECNVNS-UHFFFAOYSA-N
Synonym benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
PubChem CID 7045
ChEBI CHEBI:36616
IUPAC Name quinoxaline
SMILES C1=CC=C2C(=C1)N=CC=N2

Specifications

Melting Point 29°C to 34°C
Color Yellow to Brown
Boiling Point 220°C to 223°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 176
Merck Index 15, 8195
Solubility Information Solubility in water: soluble. Other solubilities: miscible in alcohol,ether,benzene,acetone
Formula Weight 130.15
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Quinoxaline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.