missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ QX 314 chloride

Product Code. 30564792
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30564792 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30564792 Supplier R&D Systems™ Supplier No. 2313/50
  • ONLINE EXCLUSIVE PRICE
    230.20 €
    204.75 € / 50mg
    Save 25.45 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Na+ channel blocker

Membrane impermeable quaternary derivative of lidocaine, a blocker of voltage-activated Na+ channels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1-4-5370
Quantity 50 mg
Product Line Tocris
Molecular Formula C11H18ClNO
Formula Weight 298.85
Purity >99%
Molecular Weight (g/mol) 215.72
Chemical Name or Material QX 314 chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.