missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,1'-Bi(2-naphthylamine), 97%

Product Code. 11485490
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485490 1 g 1g
15453307 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485490 Supplier Thermo Scientific Alfa Aesar Supplier No. H25785.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18741-85-0 | C20H16N2 | 284.36 g/mol

(R)-(+)-1,1'-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1,1′-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18741-85-0
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.36
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N

Specifications

Melting Point 242°C to 243°C
Quantity 1 g
Beilstein 5279621
Solubility Information Slightly soluble in water.
Optical Rotation +150° (c=1 in Pyridine)
Formula Weight 284.36
Percent Purity 97%
Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthylamine)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.