missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(1-Naphthyl)ethylamine, 99%

Product Code. 11458746
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458746 5 g 5g
11448746 1 g 1g
11468746 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458746 Supplier Thermo Scientific Alfa Aesar Supplier No. A16918.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3886-70-2 | C12H13N | 171.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-70-2
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00064114
InChI Key RTCUCQWIICFPOD-SECBINFHSA-N
Synonym r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine
PubChem CID 2724264
IUPAC Name (1R)-1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Density 1.06
Boiling Point 152°C to 153°C (11 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.623
Quantity 5 g
UN Number UN2922
Beilstein 2208025
Sensitivity Air sensitive
Formula Weight 171.24
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-1-(1-Naphthyl)ethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (R)-(+)-1-(1-Naphthyl)ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.