missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(1-Naphthyl)ethylamine Cinacalcet Impurity A, TRC

Product Code. 17993555
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993555 1 g 1g
17961645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17993555 Supplier Toronto Research Chemicals Supplier No. TRCN3783451G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-70-2
Molecular Formula C12 H13 N
Molecular Weight (g/mol) 171.24
Synonym 1-Naphthalenemethanamine, α-methyl-, (αR)-, 1-Naphthalenemethanamine, α-methyl-, (R)-, 1-Naphthalenemethylamine, α-methyl-, (R)-(+)- (8CI), (+)-1-(1-Naphthyl)ethylamine, (+)-1-(α-Naphthyl)ethylamine, (+)-[(R)-1-(Naphthalen-1-yl)ethyl]amine, (+)-α-(1-Naphthyl)ethylamine, (1R)-1-(1-Naphthyl)ethanamine, (R)-(+)-1-(1-Naphthyl)ethylamine, (R)-(+)-1-(1-Naphthyl)ethylamine, (R)-(+)-1-(α-Naphthyl)ethylamine, (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(1-Naphthalenyl)ethylamine, (R)-1-(1-Naphthyl)ethanamine, (R)-1-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (R)-1-(α-Naphthyl)ethylamine, (R)-1-Naphthylethylamine, (R)-α-(1-Naphthyl)ethylamine, (R)-α-Methyl-1-naphthalenemethanamine, (αR)-α-Methyl-1-naphthalenemethanamine, [(R)-1-(Naphthalen-1-yl)ethyl]amine, d-1-(1-Naphthyl)ethylamine
IUPAC Name (1R)-1-naphthalen-1-ylethanamine
SMILES C[C@@H](N)c1cccc2ccccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3/t9-/m1/s1
Formula Weight 171.1048
Chemical Name or Material (1R)-1-(1-Naphthyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.