missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]isoquinoline, TRC

Product Code. 30427892
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427892 100 mg -
30419085 250 mg -
30423435 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427892 Supplier Toronto Research Chemicals Supplier No. TRCO303565100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30356-08-2
Molecular Formula C17H23NO
Molecular Weight (g/mol) 257.37
Synonym (1R)-1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]isoquinoline
IUPAC Name (1S)-1-[(4-methoxyphenyl)methyl]-1,2,3,4,5,6,7,8-octahydroisoquinoline
SMILES COc1ccc(C[C@@H]2NCCC3=C2CCCC3)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H23NO/c1-19-15-8-6-13(7-9-15)12-17-16-5-3-2-4-14(16)10-11-18-17/h6-9,17-18H,2-5,10-12H2,1H3/t17-/m0/s1
Formula Weight 257.178
Chemical Name or Material (R)-1,2,3,4,5,6,7,8-Octahydro-1-[(4-methoxyphenyl)methyl]isoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.