missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid, MedChemExpress

Product Code. 16416386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16416386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16416386 Supplier Medchem Express Supplier No. HY13987
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid is a constrained Phe analogue which can fold into a beta-bend and a helical structure, and to adopt a preferred side-chain disposition in the peptide.

(R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid is a constrained Phe analogue which can fold into a beta-bend and a helical structure, and to adopt a preferred side-chain disposition in the peptide.

Specifications

Chemical Name or Material (R)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylic acid
CAS 103733-65-9
Color White
Purity Grade Notes Research
Molecular Formula C10H11NO2
Quantity 100 mg
Synonym D-phenylalanine analogue
Solubility Information H2O : 1 mg/mL (5.64 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES O=C([C@@H]1NCC2=C(C=CC=C2)C1)O
Molecular Weight (g/mol) 177.2
Formula Weight 177.2
Percent Purity 99.28%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.