missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2-Epoxybutane, 98%

Product Code. 15494818
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494818 1 g 1g
15404828 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494818 Supplier Thermo Scientific Alfa Aesar Supplier No. H58579.03
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3760-95-0 | C4H8O | 72.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3760-95-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD02683444
InChI Key RBACIKXCRWGCBB-SCSAIBSYSA-N
Synonym r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
PubChem CID 11147670
IUPAC Name (2R)-2-ethyloxirane
SMILES CCC1CO1

Specifications

Density 0.83
Boiling Point 60°C to 64°C
Flash Point −12°C (10°F)
Refractive Index 1.381
Quantity 1 g
UN Number UN3022
Optical Rotation +10° (Neat)
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material (R)-(+)-1,2-Epoxybutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.