missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 99%, ee 98+%

Product Code. 11337329
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11337329

Brand: Thermo Scientific Alfa Aesar H27461.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17061-53-9 | C8H10ClN | 155.625 g/mol

(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17061-53-9
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD06761822
InChI Key DQEYVZASLGNODG-ZCFIWIBFSA-N
Synonym r-1-3-chlorophenyl ethanamine, 1r-1-3-chlorophenyl ethanamine, r-1-3-chlorophenyl ethylamine, r-3-chloro-, a-methylbenzylamine, 1r-1-3-chlorophenyl ethan-1-amine, r-3-chloro-methylbenzylamine, r-+-1-3-chlorophenyl ethylamine, r-1-3-chlorophenyl ethylamine, chipros, r-alpha-methyl-3-chlorobenzenemethaneamine, benzenemethanamine,3-chloro-a-methyl-, ar
PubChem CID 2507697
IUPAC Name (1R)-1-(3-chlorophenyl)ethanamine
SMILES CC(C1=CC(=CC=C1)Cl)N

Specifications

Boiling Point 112°C (13 mmHg)
Quantity 5 g
UN Number UN2735
Solubility Information Insoluble in water.
Formula Weight 155.63
Percent Purity 99%
Assay ee ≥98%
Chemical Name or Material (R)-1-(3-Chlorophenyl)ethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.