missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

Product Code. 11319665
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319665 5 g 5g
11309665 1 g 1g
11329665 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11319665 Supplier Thermo Scientific Alfa Aesar Supplier No. L19119.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27298-99-3 | C8H10ClN | 155.625 g/mol

(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

Solubility
Soluble in DMSO, insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27298-99-3
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00671639
InChI Key PINPOEWMCLFRRB-ZCFIWIBFSA-N
Synonym r-1-4-chlorophenyl ethylamine, r-1-4-chlorophenyl ethanamine, 1r-1-4-chlorophenyl ethanamine, r-+-1-4-chlorophenyl ethylamine, 1r-1-4-chlorophenyl ethan-1-amine, benzenemethanamine, 4-chloro-alpha-methyl-, alphar, pubchem15222, r-4-chloro-, a-methylbenzylamine, 1r-1-4-chlorophenyl ethylamine, r-1-4-chloro-phenyl ethylamine
PubChem CID 1715226
IUPAC Name (1R)-1-(4-chlorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Cl)N

Specifications

Density 1.11
Quantity 5 g
Refractive Index 1.5418
UN Number UN2922
Beilstein 2412317
Boiling Point 232°C
Solubility Information Soluble in DMSO,insoluble in water.
Formula Weight 155.63
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-(4-Chlorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.