missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%

Product Code. 15429297
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15429297 250 mg 250mg
15419297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429297 Supplier Thermo Scientific Alfa Aesar Supplier No. H52554.MD
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 96193-27-0 | C7H14N2 | 126.203 g/mol

(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96193-27-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD09752820
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym r-octahydropyrrolo 1,2-a pyrazine, r-1,4-diazabicyclo 4.3.0 nonane, 8ar-octahydropyrrolo 1,2-a pyrazine, r-octahydro-pyrrolo 1,2-a pyrazine, 8ar-octahydropyrrolo 1,2-a piperazine, r-1,4-diazobicyclo 4.3.0 nonane, 6r-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8ar-9ci, pyrrolo 1,2-a pyrazine, octahydro-, r, pyrrolo 1,2-a pyrazine, octahydro-, 8ar
PubChem CID 7004239
IUPAC Name (8aR)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
SMILES C1CC2CNCCN2C1

Specifications

Boiling Point 65°C to 69°C (3 mmHg)
Quantity 250 mg
UN Number UN3267
Solubility Information Slightly soluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material (R)-1,4-Diazabicyclo[4.3.0]nonane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.