missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 95+%

Product Code. 10647241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647241 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10647241 Supplier Thermo Scientific Acros Supplier No. 277140010
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 72748-99-3 | C6H14N2O | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72748-99-3
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 42°C (1.8 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14N2O
Refractive Index 1.4635 to 1.4655
MDL Number MFCD00010622
Quantity 1 g
Fieser 17,15
Specific Gravity 0.97
Specific Rotation + 81.5
Synonym r-+-1-amino-2-methoxymethyl pyrrolidine, r-2-methoxymethyl pyrrolidin-1-amine, 1-pyrrolidinamine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl pyrrolidin-1-amine, r-1-amino-2-methoxymethyl pyrrolidine, r-1-+ amino-2-methoxymethylpyrrolidine, 2-methoxymethyl-1-pyrrolidinamine #, pubchem8114, r-1-amino-2-methoxymethylpyrrolidine, 2r-2-methoxymethyl pyrrolidinylamine
InChI Key BWSIKGOGLDNQBZ-ZCFIWIBFSA-N
SMILES COCC1CCCN1N
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidin-1-amine
Molecular Weight (g/mol) 130.19
PubChem CID 2724538
Formula Weight 130.19
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.