missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Amino-2-propanol, 98+%

Product Code. 10318260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10318260 1 g Glass bottle 1g
10256270 5 g Glass bottle 5g
10732495 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10318260 Supplier Thermo Scientific Acros Supplier No. 218970010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2799-16-8 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-16-8
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064428
InChI Key HXKKHQJGJAFBHI-GSVOUGTGSA-N
Synonym r---1-amino-2-propanol, r-1-aminopropan-2-ol, r-1-amino-2-propanol, 2r-1-aminopropan-2-ol, r---isopropanolamine, 2-propanol, 1-amino-, 2r, 2r---hydroxypropylamine, r---1-aminopropan-2-ol, 2r---2-hydroxypropylamine
PubChem CID 439938
ChEBI CHEBI:15675
IUPAC Name (2R)-1-aminopropan-2-ol
SMILES C[C@@H](O)CN

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 23°C to 26°C
Color Colorless to Yellow
Density 0.9540g/mL
Boiling Point 160°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2NH2
Refractive Index 1.4465 to 1.4495
Quantity 1 g
Beilstein 04, II, 736
Specific Gravity 0.954
Specific Rotation -18
Solubility Information Solubility in water: soluble
Formula Weight 75.11
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material (R)-(-)-1-Amino-2-propanol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-1-Amino-2-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.