missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Amino-2-propanol, 98%, Thermo Scientific Chemicals

Product Code. 15438999
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438999 5 g 5g
11318975 250 mg 250mg
11308975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438999 Supplier Thermo Scientific Alfa Aesar Supplier No. L14101.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2799-16-8 | Molecular Formula: C3H9NO | Molecular Weight: 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-16-8
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064428
InChI Key HXKKHQJGJAFBHI-GSVOUGTGSA-N
Synonym r---1-amino-2-propanol, r-1-aminopropan-2-ol, r-1-amino-2-propanol, 2r-1-aminopropan-2-ol, r---isopropanolamine, 2-propanol, 1-amino-, 2r, 2r---hydroxypropylamine, r---1-aminopropan-2-ol, 2r---2-hydroxypropylamine
PubChem CID 439938
ChEBI CHEBI:15675
IUPAC Name (2R)-1-aminopropan-2-ol
SMILES C[C@@H](O)CN

Specifications

Melting Point 24°C to 26°C
Boiling Point 160°C
Flash Point 71°C (159°F)
Quantity 5 g
UN Number UN3259
Beilstein 1718869
Optical Rotation −19° (c=1.8 in Water)
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (R)-(-)-1-Amino-2-propanol
compliance-icons
Product Identifier
  • (R)-(-)-1-Amino-2-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: (R)-(-)-1-Aminopropan-2-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.