missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Benzoyl-3-methylpiperazine hydrochloride, 99%, ee 99%, Thermo Scientific™

Product Code. 11354486
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354486 250 mg 250mg
11334486 1 g 1g
11344486 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354486 Supplier Thermo Scientific Alfa Aesar Supplier No. L20059.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-1-Benzoyl-3-methylpiperazine hydrochloride is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474010-60-1
Molecular Formula C12H17ClN2O
Molecular Weight (g/mol) 240.731
MDL Number MFCD05664046
InChI Key OMFYILLDCYROOO-HNCPQSOCSA-N
Synonym r-1-benzoyl-3-methylpiperazine hydrochloride, r-3-methylpiperazin-1-yl phenyl methanone hydrochloride, 3r-1-benzoyl-3-methylpiperazine hydrochloride, 3 r-methyl-1-benzoylpiperazine hydrochloride, 3-methylpiperazin-1-yl-phenylmethanone,hydrochloride, r-1-benzoyl-3-methylpiperazinehydrochloride, r-3-methyl-piperazine-1-yl-phenyl-methanone hydrochloride
PubChem CID 68840483
IUPAC Name [(3R)-3-methylpiperazin-1-yl]-phenylmethanone;hydrochloride
SMILES CC1CN(CCN1)C(=O)C2=CC=CC=C2.Cl

Specifications

Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 240.73
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-1-Benzoyl-3-methylpiperazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.