missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%

Product Code. 15439307
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439307 5 g 5g
15429307 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439307 Supplier Thermo Scientific Alfa Aesar Supplier No. H52559.06
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140695-85-8 | C11H21NO3 | 215.293 g/mol

(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140695-85-8
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
MDL Number MFCD02683202
InChI Key OJCLHERKFHHUTB-SECBINFHSA-N
Synonym r-1-boc-3-hydroxymethyl piperidine, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, r-1-boc-3-hyroxymethyl piperidine, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r---1-boc-3-hydroxymethyl piperidine
PubChem CID 1514447
IUPAC Name tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

Melting Point 91°C to 94°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Optical Rotation −20° (c=2 in Methanol)
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-Boc-3-(hydroxymethyl)piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.