missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-BOC-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific™

Product Code. 10862431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10862431 1 g 1g
10179779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10862431 Supplier Thermo Scientific Acros Supplier No. 436480010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 140695-85-8
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
InChI Key OJCLHERKFHHUTB-SECBINFHSA-N
Synonym r-1-boc-3-hydroxymethyl piperidine, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, r-1-boc-3-hyroxymethyl piperidine, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r---1-boc-3-hydroxymethyl piperidine
PubChem CID 1514447
IUPAC Name tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 93°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -11.2
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (R)-1-BOC-3-(Hydroxymethyl)piperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.