missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-BOC-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Product Code. 11986361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11986361 5 g Glass bottle 5g
11976361 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986361 Supplier Thermo Scientific Acros Supplier No. 436430050
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143900-43-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.26
InChI Key UIJXHKXIOCDSEB-MRVPVSSYSA-N
Synonym r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine
PubChem CID 1514398
IUPAC Name tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40.0°C to 42.0°C
Color Tan to White
Flash Point 104.4°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -8.5° to -12.0° (20°C, 589nm) (c=1 in CHCl3)
Formula Weight 201.26
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material (R)-1-BOC-3-Hydroxypiperidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-1-BOC-3-Hydroxypiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.