missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Cyclohexylethyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 15499109
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499109 Supplier Thermo Scientific Alfa Aesar Supplier No. L19976.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 196402-21-8
Density 1
Molecular Formula C9H15NS
Refractive Index 1.5264
MDL Number MFCD04039328
Quantity 1 g
UN Number UN2922
Sensitivity Moisture sensitive
Synonym r---1-cyclohexylethyl isothiocyanate, 1r-1-isothiocyanatoethyl cyclohexane, cyclohexane, 1r-1-isothiocyanatoethyl
InChI Key KMMWHCFHXACMCZ-MRVPVSSYSA-N
SMILES CC(C1CCCCC1)N=C=S
IUPAC Name [(1R)-1-isothiocyanatoethyl]cyclohexane
Molecular Weight (g/mol) 169.286
PubChem CID 40424844
Formula Weight 169.29
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-Cyclohexylethyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.