missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%

Product Code. 11365525
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365525 250 mg 250mg
11345525 1 g 1g
11355525 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365525 Supplier Thermo Scientific Alfa Aesar Supplier No. L19694.MD
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 104641-60-3 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104641-60-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788747
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym 3r-1-methylpyrrolidin-3-ol, r---1-methyl-3-pyrrolidinol, r-3-hydroxy-1-methyl-pyrrolidine, r-1-methyl-3-pyrrolidinol, r-1-methylpyrrolidin-3-ol, r-3-hydroxy-1-methylpyrrolidine, r---1-methyl-3-hydroxypyrrolidine, 3-pyrrolidinol, 1-methyl-, 3r, r-1-methyl-pyrrolidin-3-ol, unii-6hvw4ew50x
PubChem CID 6951332
SMILES CN1CCC(O)C1

Specifications

Density 0.92
Boiling Point 103°C to 106°C (35 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.464
Quantity 250 mg
Beilstein 4349393
Sensitivity Air sensitive; Hygroscopic
IUPAC Name (3R)-1-methylpyrrolidin-3-ol
Formula Weight 101.15
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(-)-1-Methyl-3-hydroxypyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.