missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-phenylpropylamine, 98%

Product Code. 11945251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11945251 5 g 5g
10722212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945251 Supplier Thermo Scientific Acros Supplier No. 414760050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 937-52-0 | C10H15N | 149.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-52-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00145208
InChI Key WECUIGDEWBNQJJ-UHFFFAOYNA-N
Synonym r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamine
PubChem CID 2734033
SMILES CC(N)CCC1=CC=CC=C1

Specifications

Color Colorless
Density 0.9360g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5130 to 1.5150
Quantity 5 g
Specific Gravity 0.936
Specific Rotation Condition − 11.00 (20.00°C neat)
Specific Rotation − 11.00
IUPAC Name (2R)-4-phenylbutan-2-amine
Formula Weight 149.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methyl-3-phenylpropylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-1-Methyl-3-phenylpropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.