missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-pyrrolidinol, 99%

Product Code. 11976671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976671 5 g 5g
11966671 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976671 Supplier Thermo Scientific Acros Supplier No. 439440050
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 104641-60-3 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104641-60-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788747
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym 3r-1-methylpyrrolidin-3-ol, r---1-methyl-3-pyrrolidinol, r-3-hydroxy-1-methyl-pyrrolidine, r-1-methyl-3-pyrrolidinol, r-1-methylpyrrolidin-3-ol, r-3-hydroxy-1-methylpyrrolidine, r---1-methyl-3-hydroxypyrrolidine, 3-pyrrolidinol, 1-methyl-, 3r, r-1-methyl-pyrrolidin-3-ol, unii-6hvw4ew50x
PubChem CID 6951332
IUPAC Name (3R)-1-methylpyrrolidin-3-ol
SMILES CN1CCC(O)C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Colorless
Density 0.9210g/mL
Boiling Point 50°C to 52°C (1.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.464
Quantity 5 g
Specific Gravity 0.921
Specific Rotation -7
Formula Weight 101.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-Methyl-3-pyrrolidinol
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(-)-1-Methyl-3-pyrrolidinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.