missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%

Product Code. 11349655
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349655 5 g 5g
11359655 25 g 25g
11369655 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349655 Supplier Thermo Scientific Alfa Aesar Supplier No. L19117.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3886-69-9 | C8H11N | 121.183 g/mol

(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Air Sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3886-69-9
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064405
InChI Key RQEUFEKYXDPUSK-SSDOTTSWSA-N
Synonym r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
PubChem CID 643189
ChEBI CHEBI:35322
IUPAC Name (1R)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10°C
Density 0.948
Boiling Point 187°C
Flash Point 71°C (159°F)
Odor Amine-like
Refractive Index 1.526
Quantity 5 g
UN Number UN2735
Beilstein 2410916
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Miscible with water and chloroform.
Optical Rotation +30° (c=10 in Ethanol)
Formula Weight 121.18
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(+)-1-Phenylethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.