missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress R-1479

Product Code. 16476687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476687 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476687 Supplier MedChemExpress Supplier No. HY10444

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress R-1479 (4'-Azidocytidine), a nucleoside analogue, is a specific inhibitor of RNA-dependent RNA polymerase (RdRp) of HCV. R-1479 inhibits HCV replication in the HCV subgenomic replicon system (IC50=1.28 μM).

Specifications

Molecular Weight (g/mol) 284.23 g/mol
Product Type Reference compound
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.66%
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (351.83 mM∣Need ultrasonic)
For Use With (Application) COVID-19-anti-virus
CAS 478182-28-4
Synonym 4'-Azidocytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.