missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-1-phenylethanol, 97%, ee 98%

Product Code. 11358696
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358696 1 g 1g
11368696 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358696 Supplier Thermo Scientific Alfa Aesar Supplier No. L19403.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2549-14-6 | C8H11NO | 137.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2549-14-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00239406
InChI Key ULSIYEODSMZIPX-QMMMGPOBSA-N
Synonym r-2-amino-1-phenylethanol, r---2-amino-1-phenylethanol, 1r-2-amino-1-phenylethanol, r-2-amino-1-phenylethan-1-ol, r-, a-aminomethyl benzyl alcohol, r---alpha-aminomethyl benzyl alcohol, pubchem14747, rarechem al bw 2319, chembl19363, r-2-amino-1-phenyl-ethanol
PubChem CID 6951165
IUPAC Name (1R)-2-amino-1-phenylethanol
SMILES C1=CC=C(C=C1)C(CN)O

Specifications

Melting Point 57°C to 63°C
Boiling Point 160°C (17 mmHg)
Odor Amine-like
Quantity 1 g
UN Number UN3259
Beilstein 3196196
Sensitivity Air Sensitive
Optical Rotation −43° (c=2 in Ethanol)
Formula Weight 137.18
Percent Purity 97%
Assay ee 98%
Chemical Name or Material (R)-(-)-2-Amino-1-phenylethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.