missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Amino-5-phosphonopentanoic acid, 99%, Thermo Scientific Chemicals

Product Code. 15436629
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436629 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15436629 Supplier Thermo Scientific Alfa Aesar Supplier No. J64887.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A competitive NMDA antagonist

A competitive NMDA antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79055-68-8
Molecular Formula C5H12NO5P
Molecular Weight (g/mol) 197.127
MDL Number MFCD00078839
InChI Key VOROEQBFPPIACJ-SCSAIBSYSA-N
Synonym d-ap5, 5-phosphono-d-norvaline, d-norvaline, 5-phosphono, d-apv, 2r-2-amino-5-phosphonopentanoic acid, d-2-amino-5-phosphopentanoic acid, d---2-amino-5-phosphonopentanoic acid, d--apv, d-2-amino-5-phosphonovaleric acid, d--ap-5
PubChem CID 135342
IUPAC Name (2R)-2-amino-5-phosphonopentanoic acid
SMILES C(CC(C(=O)O)N)CP(=O)(O)O

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Optical Rotation −22° (c=1 in 6N HCl)
Formula Weight 197.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (R)-(-)-2-Amino-5-phosphonopentanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.