missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine, 95%

Product Code. 15424088
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424088 250 mg 250mg
15414088 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424088 Supplier Thermo Scientific Alfa Aesar Supplier No. H57638.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22795-97-7 | C7H16N2 | 128.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22795-97-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD00671482
InChI Key UNRBEYYLYRXYCG-SSDOTTSWSA-N
Synonym r-1-ethylpyrrolidin-2-yl methanamine, r-+-2-aminomethyl-1-ethylpyrrolidine, r-2-aminomethyl-1-ethylpyrrolidine, 2r-1-ethylpyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 1-ethyl-, 2r, 2r-1-ethyl-2-pyrrolidinemethanamine, 1-ethyl-2-pyrrolidinyl methanamine #, pubchem11067, r-+-2-aminomethyl-n-ethylpyrrolidine
PubChem CID 854138
IUPAC Name [(2R)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

Density 0.929
Boiling Point 50°C to 54°C
Flash Point 57°C (135°F)
Refractive Index 1.467
Quantity 250 mg
UN Number UN1993
Sensitivity Hygroscopic
Optical Rotation +95° (c=1 in Methanol)
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material (R)-(+)-2-Aminomethyl-1-ethylpyrrolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.