missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Aminononane, ChiPros 99+%, ee 98+%, Thermo Scientific Chemicals

Product Code. 11394216
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394216 1 g 1g
11304226 5 g 5g
11314226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394216 Supplier Thermo Scientific Alfa Aesar Supplier No. L19310.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74069-74-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD03844739
InChI Key ALXIFCUEJWCQQL-SECBINFHSA-N
Synonym r-2-aminononane, r---2-aminononane, 2r-nonan-2-amine, r-2-nonanamine, r-2-nonylamine, 2-nonanamine, 2r, r-2-aminononane, chipros r , produced by basf
PubChem CID 22831496
IUPAC Name (2R)-nonan-2-amine
SMILES CCCCCCCC(C)N

Specifications

Density 0.782
Boiling Point 73°C (19 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.4271
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Optical Rotation −6° (Neat)
Formula Weight 143.27
Percent Purity ≥99%
Assay ee ≥98%
Chemical Name or Material (R)-(-)-2-Aminononane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.