missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-(Benzyloxy)-4-((5-oxotetrahydrofuran-2-yl)methyl)phenyl Neopentyl Sulfate, TRC

Product Code. 30343124
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343124 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343124 Supplier Toronto Research Chemicals Supplier No. TRCB28671025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-2-(Benzyloxy)-4-((5-oxotetrahydrofuran-2-yl)methyl)phenyl Neopentyl Sulfate
CAS 2518203-29-5
Molecular Formula C23 H28 O7 S
Quantity 25 mg
InChI Formula InChI=1S/C23H28O7S/c1-23(2,3)16-28-31(25,26)30-20-11-9-18(13-19-10-12-22(24)29-19)14-21(20)27-15-17-7-5-4-6-8-17/h4-9,11,14,19H,10,12-13,15-16H2,1-3H3/t19-/m0/s1
SMILES CC(C)(C)COS(=O)(=O)Oc1ccc(C[C@@H]2CCC(=O)O2)cc1OCc3ccccc3
IUPAC Name 2,2-dimethylpropyl [4-[[(2S)-5-oxooxolan-2-yl]methyl]-2-phenylmethoxyphenyl] sulfate
Molecular Weight (g/mol) 448.529
Formula Weight 448.156
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.