Learn More
Tocris Bioscience™ (R)-2-Hydroxyglutaric acid disodium salt, Tocris Bioscience™
GREENER_CHOICE
KDM4A inhibitor; also NF-κB activator
Brand:  Tocris Bioscience™ 6124/50
Encompass
Product Code. 15757226
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
(R)-2-Hydroxyglutaric acid disodium salt is an oncometabolite produced by mutant isocitrate dehydrogenase 1 (IDH1) and IDH2. Inhibits KDM4A in murine embryonic fibroblasts resulting in activation of mTOR. Also activates NF-κB in bone marrow stromal cells and inhibits FTO (mRNA N6-methyladenine demethylase).
Specifications
Specifications
(R)-2-Hydroxyglutaric acid disodium salt
(R)-2-Hydroxypentanedioic acid disodium salt
Soluble to 100mM in water
White
103404-90-6
50 mg
Solid
Bioactive Small Molecule