missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Hydroxymethylmorpholine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15411019
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15411019 1 g 1g
15428636 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15411019 Supplier Thermo Scientific Alfa Aesar Supplier No. H64003.03
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436436-17-7
Molecular Formula C5H12ClNO2
MDL Number MFCD11052541
Synonym r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethylmorpholine hcl, r-morpholin-2-yl methanol hydrochloride, r-2-hydroxymethylmorpholine hydrochloride, r-morpholin-2-ylmethanol-hcl, r-2-morpholinemethanol hcl, r-2-morpholinemethanol hydrochloride, 2r-morpholin-2-ylmethanol hydrochloride, r-2-hydroxymethyl-morpholine hydrochloride, r-2-hydroxymethyl morpholine hydrochloride
PubChem CID 42614735
IUPAC Name [(2R)-morpholin-2-yl]methanol;hydrochloride

Specifications

Quantity 1 g
InChI Key NBGXGDBTUJNTKJ-NUBCRITNSA-N
SMILES C1COC(CN1)CO.Cl
Molecular Weight (g/mol) 153.606
Formula Weight 153.61
Percent Purity 95%
Physical Form Solid
Chemical Name or Material (R)-2-Hydroxymethylmorpholine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.