missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%

Product Code. 10862641 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10862641 5 mL 5mL
10200800 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10862641 Supplier Thermo Scientific Supplier No. 376190050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84025-81-0 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84025-81-0
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key CHPRFKYDQRKRRK-ZCFIWIBFSA-N
Synonym r-2-methoxymethyl pyrrolidine, 2r-2-methoxymethyl pyrrolidine, r---2-methoxymethyl pyrrolidine, o-methyl-d-prolinol, r-2-methoxymethyl-pyrrolidine, r-2-methoxymethylpyrrolidine, pyrrolidine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl-pyrrolidine, r-+-2-methoxymethyl pyrrolidine, h-pro-ol-me
PubChem CID 671216
IUPAC Name (2R)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9320g/mL
Boiling Point 62.0°C (40.0 mmHg)
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 mL
Specific Gravity 0.932
Specific Rotation -32.5° to -36.5° (20°C, 589nm) (c=2 in aq HCl)
Solubility Information Solubility in water: soluble
Formula Weight 115.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (R)-(-)-2-(Methoxymethyl)pyrrolidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (R)-(-)-2-(Methoxymethyl)pyrrolidine, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.