missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Phenyl-1-propanol, 98+%

Product Code. 11328875
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328875 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11328875 Supplier Thermo Scientific Alfa Aesar Supplier No. L13999.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19141-40-3 | C9H12O | 136.19 g/mol

For synthesis of optically active products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19141-40-3
Density 0.975
Boiling Point 220°C
Flash Point 108°C (226°F)
Molecular Formula C9H12O
Refractive Index 1.526
MDL Number MFCD00145206
Quantity 250 mg
Synonym r-2-phenyl-1-propanol, r-+-2-phenyl-1-propanol, 2r-2-phenylpropan-1-ol, r-2-phenylpropanol, unii-9d7u4y35ur, 2-phenyl-1-propanol, +-, 2r-2-phenyl-1-propanol, +--2-phenyl-1-propanol, benzeneethanol, beta-methyl-, r, r-2-phenyl-1-propanol, +-
InChI Key RNDNSYIPLPAXAZ-QMMMGPOBSA-N
Optical Rotation +18° (Neat)
SMILES C[C@@H](CO)C1=CC=CC=C1
IUPAC Name (2R)-2-phenylpropan-1-ol
Molecular Weight (g/mol) 136.19
PubChem CID 177050
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-2-Phenyl-1-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.